missing translation for 'onlineSavingsMsg'
Learn More

NSC16168, MedChemExpress

Product Code. 16445857
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16445857

Brand: Medchem Express HY100690

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NSC16168 is a specific inhibitor of ERCC1-XPF, with an IC50 value of 0.42 μM. NSC16168 inhibits DNA repair and potentiates CDDP efficacy in cancer.

NSC16168 is a specific inhibitor of ERCC1-XPF, with an IC50 value of 0.42 μM. NSC16168 inhibits DNA repair and potentiates CDDP efficacy in cancer.

Specifications

Chemical Name or Material NSC16168
CAS 6837-93-0
Color Light Nude
Purity Grade Notes Research
Molecular Formula C17H15NO9S3
Quantity 5 mg
Solubility Information DMSO : 31.25 mg/mL (66.00 mM; Need ultrasonic)
SMILES NC1=C2C(C=C(S(=O)(O)=O)C=C2OS(C3=CC=C(C)C=C3)(=O)=O)=CC(S(O)(=O)=O)=C1
Molecular Weight (g/mol) 473.5
Formula Weight 473.5
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.