missing translation for 'onlineSavingsMsg'
Learn More

NSC 74859, MedChemExpress

Product Code. 16498907
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498907 5 mg 5mg
16492437 50 mg 50mg
16436328 10 mg 10mg
16436646 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498907 Supplier Medchem Express Supplier No. HY15146
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress NSC 74859 (S3I-201) is a selective Stat3 inhibitor with an IC50 of 86 μM.

NSC 74859 (S3I-201) is a selective Stat3 inhibitor with an IC50 of 86 μM.

Specifications

Chemical Name or Material NSC 74859
CAS 501919-59-1
Color Gray
Purity Grade Notes Research
Molecular Formula C16H15NO7S
Quantity 5 mg
Synonym S3I-201
Solubility Information DMSO : 100 mg/mL (273.70 mM; Need ultrasonic)
SMILES OC1=CC(NC(COS(=O)(C2=CC=C(C=C2)C)=O)=O)=CC=C1C(O)=O
Molecular Weight (g/mol) 365.36
Formula Weight 365.36
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.