missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NSC 668036
GREENER_CHOICE

Product Code. p-7121011
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18766159

Brand: Tocris Bioscience™ 5813/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Wnt signaling inhibitor; binds the PDZ domain of the Dishevelled (Dvl) protein

NSC 668036 is a Wnt signaling inhibitor. Binds the PDZ domain of the Dishevelled (Dvl) protein. Inhibits proliferation, migration and TGF-β-induced differentiation of fibroblasts in vitro. Reduces severity of pulmonary fibrosis in a mouse model.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(1,1-dimethylethoxy)carbonyl]-L-alanyl-(2S)-2-hydroxy-3-methylbutanoyl-L-alanine-(1S)-1-carboxy-2-methylpropyl ester
CAS 144678-63-7
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Molecular Formula C21H36N2O9
Formula Weight 460.52
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.