missing translation for 'onlineSavingsMsg'
Learn More

NSC 405020, MedChemExpress

Product Code. 16492917
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16492917

Brand: Medchem Express HY15827

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NSC 405020 is a novel small molecule inhibitor of MT1-MMP that specifically targets PEX domain rather than the catalytic domain of MT1-MMP with IC50 >100 μM and does not inhibit the catalytic activity of MT1-MMP or MMP-2.

NSC 405020 is a novel small molecule inhibitor of MT1-MMP that specifically targets PEX domain rather than the catalytic domain of MT1-MMP with IC50 >100 μM and does not inhibit the catalytic activity of MT1-MMP or MMP-2.

Specifications

Chemical Name or Material NSC 405020
Physical Form Solid
CAS 7497-07-6
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C12H15Cl2NO
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 260 mg/mL (999.38 mM)
SMILES O=C(NC(C)CCC)C1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol) 260.16
Formula Weight 260.16
Percent Purity 99.75%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.