Learn More
Tocris Bioscience™ NSC 3852
Histone deacetylase inhibitor
Specifications
Purity >95%
View More Specs
Description
Description
Histone deacetylase inhibitor. Causes cell differentiation and antiproliferative activity in MCF-7 human breast cancer cells in vitro and displays antitumor activity in vivo.
Specifications
Specifications
>95%