missing translation for 'onlineSavingsMsg'
Learn More

NSC 379317, TRC

Product Code. 30420804
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30420804 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420804 Supplier Toronto Research Chemicals Supplier No. TRCN900295100MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-[((Carboxymethyl)imino)diethylene]di-(2,6-morpholinedione)
CAS 23911-26-4
Molecular Formula C14H19N3O8
Quantity 100 mg
Synonym N,N-Bis[2-(2,6-dioxo-4-morpholinyl)ethyl]glycine, 1,5-Bis(2,6-dioxomorpholino)-3-azapentane-3-acetic Acid, 2-[Bis[2-(2,6-dioxomorpholino)ethyl]amino]acetic Acid, Cyclic DTPA Anhydride, DTPA Anhydride, DTPA Ayclic Anhydride, DTPA Dianhydride, DTPAA, Diethylenetriamine-N,N,N',N'',N''-pentaacetic Acid N,N''-Dianhydride, Diethylenetriamine-N,N,N',N'',N''-pentacetic Dianhydride, Diethylenetriaminepentaacetic Acid Dianhydride, Diethylenetriaminepentaacetic Bisanhydride, Diethylenetriaminepentaacetic Dianhydride, NSC 379317
InChI Formula InChI=1S/C14H19N3O8/c18-10(19)5-15(1-3-16-6-11(20)24-12(21)7-16)2-4-17-8-13(22)25-14(23)9-17/h1-9H2,(H,18,19)
SMILES OC(=O)CN(CCN1CC(=O)OC(=O)C1)CCN2CC(=O)OC(=O)C2
IUPAC Name 2-[bis[2-(2,6-dioxomorpholin-4-yl)ethyl]amino]acetic acid
Molecular Weight (g/mol) 357.32
Formula Weight 357.1172
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.