missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NSC 23766

Product Code. p-5545931
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18765346

Brand: Tocris Bioscience™ 2161/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective inhibitor of Rac1-GEF interaction; antioncogenic

NSC 23766 is a selective inhibitor of Rac1-GEF interaction. NSC 23766 prevents Rac1 activation by Rac-specific guanine nucleotide exchange factors (GEFs) TrioN and Tiam1 (IC50 ∼ 50 μM) without affecting Cdc42 or RhoA activation. The compound inhibits Rac1-mediated cell functions and is reported to reverse tumor cell phenotypes in prostate cancer cells. NSC 23766 also inhibits mammary carcinoma cell invasion in vitro and inhibits mammary tumor progression and metastasis in a mouse model.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type NSC 23766
Purity >96%
Molecular Weight (g/mol) 530.97
Inhibitors Rho GTPase Inhibitors

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.