missing translation for 'onlineSavingsMsg'
Learn More

NSC 228155, MedChemExpress

Product Code. 16499796
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499796 50 mg 50mg
16495977 5 mg 5mg
16484516 100 mg 100mg
16443258 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499796 Supplier Medchem Express Supplier No. HY101084

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NSC 228155 is an activator of EGFR, binds to the extracellular region of EGFR and enhance tyrosine phosphorylation of EGFR. NSC 228155 is also a potent inhibitor of KIX-KID interaction, inhibits kinase-inducible domain (KID) from CREB and KID-interacting domain (KIX) from CBP, with an IC50 of 0.36 μM.

NSC 228155 is an activator of EGFR, binds to the extracellular region of EGFR and enhance tyrosine phosphorylation of EGFR. NSC 228155 is also a potent inhibitor of KIX-KID interaction, inhibits kinase-inducible domain (KID) from CREB and KID-interacting domain (KIX) from CBP, with an IC50 of 0.36 μM.

Specifications

Chemical Name or Material NSC 228155
Physical Form Solid
CAS 113104-25-9
Color Orange
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C11H6N4O4S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 16 mg/mL (55.12 mM; Need ultrasonic)
SMILES [O-][N+]1=CC=CC=C1SC2=CC=C([N+]([O-])=O)C3=NON=C32
Molecular Weight (g/mol) 290.25
Formula Weight 290.25
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.