missing translation for 'onlineSavingsMsg'
Learn More

NSC 185058, MedChemExpress

Product Code. 16497947
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497947 5 mg 5mg
16495228 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16497947 Supplier Medchem Express Supplier No. HY125169
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NSC 185058 is an inhibitor of ATG4B, a major cysteine protease. Inhibition of ATG4B using NSC 185058 markedly attenuates autophagic activity.

NSC 185058 is an inhibitor of ATG4B, a major cysteine protease. Inhibition of ATG4B using NSC 185058 markedly attenuates autophagic activity.

Specifications

Chemical Name or Material NSC 185058
CAS 39122-38-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C11H9N3S
Quantity 5 mg
Solubility Information DMSO : ≥ 125 mg/mL (580.67 mM)
SMILES S=C(C1=NC=CC=C1)NC2=NC=CC=C2
Molecular Weight (g/mol) 215.27
Formula Weight 215.27
Percent Purity 99.52%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.