missing translation for 'onlineSavingsMsg'
Learn More

NS309, MedChemExpress

Product Code. 16429087
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16429087

Brand: Medchem Express HY15416

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NS309 is a potent and selective activator of the Ca2+-activated SK/IK potassium channels, but displays no activity at BK channels.

NS309 is a potent and selective activator of the Ca2+-activated SK/IK potassium channels, but displays no activity at BK channels.

Specifications

Chemical Name or Material NS309
Physical Form Solid
CAS 18711-16-5
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C8H4Cl2N2O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (432.83 mM; Need ultrasonic)
SMILES O=C1NC2=C(C=CC(Cl)=C2Cl)/C1=N/O
Molecular Weight (g/mol) 231.04
Formula Weight 231.04
Percent Purity 99.19%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.