missing translation for 'onlineSavingsMsg'
Learn More

NS1652, MedChemExpress

Product Code. 16450559
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16450559

Brand: Medchem Express HY100244

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NS1652 is a reversible anion conductance inhibitor, blocks chloride channel, with an IC50 of 1.6 μM in human and mouse red blood cells.

NS1652 is a reversible anion conductance inhibitor, blocks chloride channel, with an IC50 of 1.6 μM in human and mouse red blood cells.

Specifications

Chemical Name or Material NS1652
Physical Form Solid
CAS 1566-81-0
Color White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C15H11F3N2O3
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 5 mg/mL (15.42 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=CC=C1NC(NC2=CC=CC(C(F)(F)F)=C2)=O
Molecular Weight (g/mol) 324.25
Formula Weight 324.25
Percent Purity 99.89%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.