missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NS11394, MedChemExpress
Description
NS11394 is an orally active and unique subtype-selective GABAA positive allosteric receptor (PAM), with a Ki of ∼0.5 nM. NS11394 shows a selectivity profile in the order of GABAA-5 > α3 > α2 > α1-containing receptors. NS11394 has anxiolytic and anti-inflammatory properties.
Specifications
Specifications
| Chemical Name or Material | NS11394 |
| CAS | 951650-22-9 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H19N3O |
| Quantity | 5 mg |
| Solubility Information | DMSO : 100 mg/mL (282.95 mM; Need ultrasonic) |
| SMILES | N#CC1=CC=CC=C1C2=CC=CC(N3C4=CC=C(C(C)(O)C)C=C4N=C3)=C2 |
| Molecular Weight (g/mol) | 353.42 |
| Formula Weight | 353.42 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?