missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NS1-BP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93108-0.02ml
This item is not returnable.
View return policy
Description
NS1-BP Polyclonal antibody specifically detects NS1-BP in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NS1-BP | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| ARA3, aryl hydrocarbon receptor-associated 3, Aryl hydrocarbon receptor-associated protein 3, DKFZp686K06216, FLARA3, FLJ10962, FLJ35593, FLJ36593, HSPC068, influenza virus NS1A binding protein, influenza virus NS1A-binding protein, KIAA0850FLJ10069, KLHL39, NCX downstream gene 1, ND1, NS1, NS-1, NS1-binding protein, NS1BP, NS1-BPFLJ10411 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human IVNS1ABP (NP_006460.2). MIPNGYLMFEDENFIESSVAKLNALRKSGQFCDVRLQVCGHEMLAHRAVLACCSPYLFEIFNSDSDPHGISHVKFDDLNPEAVEVLLNYAYTAQLKADKELVKDVYSAAKKLKMDRVKQVCGDYLLSRMDVTSCISYRNFASCMGDSRLLNKVDAYIQEHLLQISEEEEFLKLPRLKLEVMLEDNVCLPSNGKLYTKVINWVQRSIWENGDSLEELMEEVQTLYYSADHKLLDGNLLDGQAEVFGSDDDHIQFVQKKPPRENGHKQISSSSTGCLSSPNATVQSPKHEWKIVASEKTSNN | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10625 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction