missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NS 9283
GREENER_CHOICE

Product Code. 18765999
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18765999

Brand: Tocris Bioscience™ 5112/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Positive allosteric modulator of α4β2 nAChRs

NS 9283 is a positive allosteric modulator of α4β2 receptor; increases the potency of Ach-evoked currents ∼60 fold without affecting the maximum efficacy (HEK293-hα4β2 cells). Reduces the rate of recovery from desensitization and slows the rate of deactivation. Displays a synergistic effect on the Emax when given with NS206 at 3α:2β receptors.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type NS 9283
Purity >99%
Molecular Weight (g/mol) 248.24
Inhibitors NS 9283

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.