missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ NS 8593 Hydrochloride

Product Code. 18753088
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18753088 50 mg 50mg
18750939 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18753088 Supplier R&D Systems™ Supplier No. 4597/50
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective negative modulator of KCa2 (SK) channels; inhibits SK currents

NS 8593 hydrochloride is a selective KCa2 (SK) channel negative modulator; inhibits SK channel currents (Kd values are 0.42, 0.6 and 0.73 μM for SK1, SK2 and SK3 respectively at 0.5 μM Ca2+). Exhibits selectivity for SK channels over KCa1.1 (BK), KCa3.1 (IK), Kv, Nav and Cav channels. Inhibits afterhyperpolarization in hippocampal slices.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material NS 8593 hydrochloride
CAS 875755-24-1
Synonym N-[(1R)-1,2,3,4-Tetrahydro-1-naphthalenyl]-1H-benzimidazol-2-amine hydrochloride
Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Product Line Tocris
Molecular Formula C17H17N3·HCl
Formula Weight 299.8
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.