Learn More
Tocris Bioscience™ NS 3763
Selective non-competitive kainate antagonist; selective for GLUK5
Specifications
Purity >98%
View More Specs
Description
Description
Selective non-competitive kainate receptor antagonist; displays selectivity for GluK1 (formerly GLUK5) subunit-containing receptors (IC50 values are 1.6 and > 30 μM for homomeric GluK1 (formerly GLUK5) and GluK2 (formerly GLUK6) receptors respectively). Has minimal activity at AMPA and NMDA receptors (IC50> 30 μM).
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
Specifications
>98%