missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NS 309, Tocris Bioscience™
Positive modulator of KCa2 and KCa3.1 channels
£109.56 - £361.88
Chemical Identifiers
CAS | 18711-16-5 |
---|---|
Molecular Formula | C8H4Cl2N2O2 |
Molecular Weight (g/mol) | 231.032 |
InChI Key | CVOUSAVHMDXCKG-UHFFFAOYSA-N |
Synonym | 6,7-dichloro-1h-indole-2,3-dione 3-oxime, 6,7-dichloro-3-hydroxyimino indolin-2-one, z-6,7-dichloro-3-hydroxyimino indolin-2-one, 6,7-dichloro-3-hydroxyamino indol-2-one, 3-oxime-6,7-dichloro-1h-indole-2,3-dione, 6,7-dichloro-3-hydroxyamino-2h-indol-2-one, 3e-6,7-dichloro-3-hydroxyimino-1,3-dihydro-2h-indol-2-one, 3e-6,7-dichloro-3-hydroxyimino-1h-indol-2-one, ns309 hplc, ns hplc , solid |
PubChem CID | 11637204 |
IUPAC Name | 6,7-dichloro-3-(hydroxyamino)indol-2-one |
SMILES | C1=CC(=C(C2=NC(=O)C(=C21)NO)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18004152
|
Tocris Bioscience
3895/10 |
10 mg |
£109.56
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18045532
|
Tocris Bioscience
3895/50 |
50 mg |
£361.88
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Positive modulator of small- and intermediate- conductance Ca2+-activated K+ channels (KCa2 and KCa3.1 channels); increases Ca2+ sensitivity. Displays no activity at BK channels.Chemical Identifiers
18711-16-5 | |
231.032 | |
6,7-dichloro-1h-indole-2,3-dione 3-oxime, 6,7-dichloro-3-hydroxyimino indolin-2-one, z-6,7-dichloro-3-hydroxyimino indolin-2-one, 6,7-dichloro-3-hydroxyamino indol-2-one, 3-oxime-6,7-dichloro-1h-indole-2,3-dione, 6,7-dichloro-3-hydroxyamino-2h-indol-2-one, 3e-6,7-dichloro-3-hydroxyimino-1,3-dihydro-2h-indol-2-one, 3e-6,7-dichloro-3-hydroxyimino-1h-indol-2-one, ns309 hplc, ns hplc , solid | |
6,7-dichloro-3-(hydroxyamino)indol-2-one |
C8H4Cl2N2O2 | |
CVOUSAVHMDXCKG-UHFFFAOYSA-N | |
11637204 | |
C1=CC(=C(C2=NC(=O)C(=C21)NO)Cl)Cl |
Specifications
18711-16-5 | |
6,7-dichloro-1h-indole-2,3-dione 3-oxime, 6,7-dichloro-3-hydroxyimino indolin-2-one, z-6,7-dichloro-3-hydroxyimino indolin-2-one, 6,7-dichloro-3-hydroxyamino indol-2-one, 3-oxime-6,7-dichloro-1h-indole-2,3-dione, 6,7-dichloro-3-hydroxyamino-2h-indol-2-one, 3e-6,7-dichloro-3-hydroxyimino-1,3-dihydro-2h-indol-2-one, 3e-6,7-dichloro-3-hydroxyimino-1h-indol-2-one, ns309 hplc, ns hplc , solid | |
C1=CC(=C(C2=NC(=O)C(=C21)NO)Cl)Cl | |
231.032 | |
231.04 | |
NS 309 |
C8H4Cl2N2O2 | |
CVOUSAVHMDXCKG-UHFFFAOYSA-N | |
6,7-dichloro-3-(hydroxyamino)indol-2-one | |
11637204 | |
>98% |
Safety and Handling
Recommended Storage : Store at +4°C