missing translation for 'onlineSavingsMsg'
Learn More

NS 1738, MedChemExpress

Product Code. 16491237
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491237 50 mg 50mg
16477727 5 mg 5mg
16475756 100 mg 100mg
16464998 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491237 Supplier Medchem Express Supplier No. HY12151
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress NS 1738 (NSC 213859) is a novel positive allosteric modulator of the α7 nAChR, with respect to positive modulation of α7 nAChR (EC50=3.4 μM in oocyte experiments).

NS 1738 (NSC 213859) is a novel positive allosteric modulator of the α7 nAChR, with respect to positive modulation of α7 nAChR (EC50=3.4 μM in oocyte experiments).

Specifications

Chemical Name or Material NS 1738
CAS 501684-93-1
Color White
Purity Grade Notes Research
Molecular Formula C14H9Cl2F3N2O2
Quantity 50 mg
Synonym NSC 213859
Solubility Information DMSO : ≥ 100 mg/mL (273.88 mM)
SMILES O=C(NC1=CC(C(F)(F)F)=CC=C1Cl)NC2=CC(Cl)=CC=C2O
Molecular Weight (g/mol) 365.13
Formula Weight 365.13
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.