missing translation for 'onlineSavingsMsg'
Learn More

NS-1619, MedChemExpress

Product Code. 16475208
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16475208

Brand: Medchem Express HY12496

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NS-1619 is an opener of large conductance Ca2+-activated K+ (BK) channel. NS-1619 is a highly effective relaxant with an EC50 of about 10 - 30 μM in several smooth muscles of blood vessels and other tissues. NS1619 inhibits proliferation and induces apoptosis in A2780 ovarian cancer cells.

NS-1619 is an opener of large conductance Ca2+-activated K+ (BK) channel. NS-1619 is a highly effective relaxant with an EC50 of about 10 - 30 μM in several smooth muscles of blood vessels and other tissues. NS1619 inhibits proliferation and induces apoptosis in A2780 ovarian cancer cells.

Specifications

Chemical Name or Material NS-1619
CAS 153587-01-0
Color White
Purity Grade Notes Research
Molecular Formula C15H8F6N2O2
Quantity 10 mg
Solubility Information DMSO : ≥ 31 mg/mL (85.58 mM)
SMILES O=C1NC2=CC(C(F)(F)F)=CC=C2N1C3=CC(C(F)(F)F)=CC=C3O
Molecular Weight (g/mol) 362.23
Formula Weight 362.23
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.