missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NS 11021

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Activator of large-conductance Ca2+-activated potassium channels (BKCa, KCa1.1). Exhibits no modulatory effect on a variety of K+ (KV), Na+ and Ca2+ currents at concentrations <10 μM. Alters gating kinetics, but does not affect single channel conductance. Shown to bind BKCa in open and closed conformations; thought to bind the α subunit.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 511.24 |
| Purity | >98% |
| Molecular Weight (g/mol) | 511.244 |
| Chemical Name or Material | NS 11021 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction