missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NRXN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549444
This item is not returnable.
View return policy
Description
NRXN1 Polyclonal Antibody for Western Blot
Neuronal cell surface protein that may be involved in cell recognition and cell adhesion by forming intracellular junctions through binding to neuroligins. May play a role in formation or maintenance of synaptic junctions. May mediate intracellular signaling. May play a role in angiogenesis.
Specifications
| NRXN1 | |
| Polyclonal | |
| Unconjugated | |
| Nrxn1 | |
| 1700062G21Rik; 9330127H16Rik; A230068P09Rik; alpha-latrotoxin receptor (calcium-dependent); DKFZp313P2036; FLJ35941; Hs.22998; Kiaa0578; mKIAA0578; neurexin 1; Neurexin 1alpha; neurexin I; neurexin I-alpha; Neurexin I-beta; neurexin-1; Neurexin-1-alpha; neurexin-1-beta; non-processed neurexin I-alpha; Nrxn1; Nrxn1b; PTHSL2; SCZD17 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 18189, 60391, 9378 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P0DI97, P58400, Q63372, Q9CS84, Q9ULB1 | |
| Nrxn1 | |
| KLH conjugated synthetic peptide between 77-110 amino acids from the N-terminal region of human NRXN1. | |
| 400 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction