missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRXN1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94557-0.02ml
This item is not returnable.
View return policy
Description
NRXN1 Polyclonal antibody specifically detects NRXN1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| NRXN1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp313P2036, FLJ35941, KIAA0578Neurexin I-alpha, neurexin 1, neurexin I, Neurexin I-beta, neurexin-1-alpha, neurexin-1-beta, PTHSL2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 31-310 of human NRXN1 (NP_001129131.1). LEFPGAEGQWTRFPKWNACCESEMSFQLKTRSARGLVLYFDDEGFCDFLELILTRGGRLQLSFSIFCAEPATLLADTPVNDGAWHSVRIRRQFRNTTLFIDQVEAKWVEVKSKRRDMTVFSGLFVGGLPPELRAAALKLTLASVREREPFKGWIRDVRVNSSQVLPVDSGEVKLDDEPPNSGGGSPCEAGEEGEGGVCLNGGVCSVVDDQAVCDCSRTGFRGKDCSQEIKFGLQCVLPVLLHDNDQGKYCCINTAKPLTEKDNNVEGLAHLMMGDQGKSK | |
| 0.02 mL | |
| Cell Biology, Neuroscience, Signal Transduction | |
| 9378 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction