missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NRX 252114
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
NRX-252114 is a potent enhancer of the interaction between S37A mutant β-catenin, and its cognate E3 ligase, SCFβ-TrCP (EC50 = 6.5 nM; Kd = 0.4 nM). NRX-252114 is a molecular glue degrader that binds at the phosphoserine binding site on β-TrCP and induces mutant β-catenin ubiquitination and subsequent degradation.
Specifications
Specifications
| Chemical Name or Material | 2-[[4-[(2,6-Dichlorophenyl)thio]-1,2-dihydro-2-oxo-6-(trifluoromethyl)-3-pyridinyl]carbonyl]-2,3-dihydro-1H-isoindole-4-carbonitrile |
| CAS | 2763260-39-3 |
| Quantity | 25 mg |
| Target | Molecular Glues |
| Molecular Formula | C22H12Cl2F3N3O2S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction