missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NRX 252114

Product Code. 30023138
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 30023138

Brand: Tocris Bioscience™ 7990/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Molecular glue Degrader of mutant β-catenin

NRX-252114 is a potent enhancer of the interaction between S37A mutant β-catenin, and its cognate E3 ligase, SCFβ-TrCP (EC50 = 6.5 nM; Kd = 0.4 nM). NRX-252114 is a molecular glue degrader that binds at the phosphoserine binding site on β-TrCP and induces mutant β-catenin ubiquitination and subsequent degradation.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[[4-[(2,6-Dichlorophenyl)thio]-1,2-dihydro-2-oxo-6-(trifluoromethyl)-3-pyridinyl]carbonyl]-2,3-dihydro-1H-isoindole-4-carbonitrile
CAS 2763260-39-3
Quantity 25 mg
Target Molecular Glues
Molecular Formula C22H12Cl2F3N3O2S
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.