missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ nrv2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532170
This item is not returnable.
View return policy
Description
Recommended positive controls: Drosophila lysate. Predicted reactivity: Drosophila (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This is the non-catalytic component of the active enzyme, which catalyzes the hydrolysis of ATP coupled with the exchange of Na(+) and K(+) ions across the plasma membrane. The beta subunit regulates, through assembly of alpha/beta heterodimers, the number of sodium pumps transported to the plasma membrane.
Specifications
| nrv2 | |
| Polyclonal | |
| Unconjugated | |
| nrv2 | |
| 42kDa; CG9261; CG9261-PA; CG9261-PC; CG9261-PD; CG9261-PE; CG9261-PF; CG9261-PG; Dmel\CG9261; Dmel_CG9261; ecta; ectatic; GH13134p; IP16413p; K-ATPase; l(2)k04223; l(2)k13315; l(2)SH0329; l(2)SH2 0329; Na; Na,K-ATPase beta-subunit; Na[+]/K[+] ATPase; Na[+]/K[+]ATPase; Na[+]/K[+]-ATPase; Na+/K+ ATPase; NaK ATPase; nervana; nervana 2; nervana2; NEST:bs13c10; nirvana; nirvana2; Nrv; nrv2; nrv2.2; nrv2-PA; nrv2-PC; nrv2-PD; nrv2-PE; nrv2-PF; nrv2-PG; Protein nervana 2; Sodium/potassium-dependent ATPase subunit beta-2; Sodium/potassium-transporting ATPase subunit beta-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 33953 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q24048 | |
| nrv2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Drosophila nrv2.1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Drosophila | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction