Learn More
NRSN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563920
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VKSYSKEEKF LQQKFKERIA DIKAHTQPVT KAPGPGETKI PVTLSRVQNV QPLLAT Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.
May play an important role in neural organelle transport, and in transduction of nerve signals or in nerve growth. May play a role in neurite extension. May play a role in memory consolidation. [UniProt]Specifications
NRSN1 | |
Polyclonal | |
Unconjugated | |
NRSN1 | |
neurensin 1; neurensin-1; Neuro-p24; nrsn1; p24; vesicular membrain protein p24; vesicular membrane protein of 24 kDa; Vesicular membrane protein p24; Vmp | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140767 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IZ57 | |
NRSN1 | |
Recombinant protein corresponding to Human NRSN1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only