missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRSF Rabbit anti-Human, Mouse, Rat, Clone: 8R8Z4, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | NRSF |
|---|---|
| Clone | 8R8Z4 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18313253
|
Bio-Techne
NBP3-16128-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18384446
|
Novus Biologicals
NBP3-16128-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NRSF Monoclonal antibody specifically detects NRSF in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| NRSF | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers, Transcription Factors and Regulators | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5978 | |
| IgG | |
| Affinity purified |
| 8R8Z4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Neural-restrictive silencer factor, NRSFneuron restrictive silencer factor, RE1-silencing transcription factor, X2 box repressor, XBRrepressor binding to the X2 box | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human NRSF (Q13127). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title