missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRSF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-757
This item is not returnable.
View return policy
Description
NRSF Polyclonal specifically detects NRSF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP).
Specifications
| NRSF | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| Neural-restrictive silencer factor, NRSFneuron restrictive silencer factor, RE1-silencing transcription factor, X2 box repressor, XBRrepressor binding to the X2 box | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
| ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:2500-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:2000, Chromatin Immunoprecipitation (ChIP) | |
| NP_005603.2 | |
| REST | |
| The immunogen recognized by this antibody maps to a region between residue 1050 and the C-terminus (residue 1097) of human RE1-silencing transcription factor using the numbering given in entry NP_005603.2 (GeneID 5978). | |
| 100 μL | |
| Neuronal Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators | |
| 5978 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction