missing translation for 'onlineSavingsMsg'
Learn More

NRSF Antibody (CL0381), Novus Biologicals™

Mouse Monoclonal Antibody

£292.00 - £436.00

Specifications

Antigen NRSF
Clone CL0381
Dilution Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 2-10 μg/ml, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18652187
View Documents
Novus Biologicals
NBP2-42048-25ul
25 μL
£292.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18628237
View Documents
Novus Biologicals
NBP2-42048
0.1 mL
£436.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NRSF Monoclonal antibody specifically detects NRSF in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

NRSF
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 2-10 μg/ml, Immunohistochemistry-Paraffin 1:50 - 1:200
Monoclonal
Purified
RUO
Human
Neural-restrictive silencer factor, NRSFneuron restrictive silencer factor, RE1-silencing transcription factor, X2 box repressor, XBRrepressor binding to the X2 box
This antibody was developed against Recombinant Protein corresponding to amino acids:SPPLPKENLREEASGDQKLLNTGEGNKEAPLQKVGAEEADESLPGLAANINESTHISSSGQNLNTPEGETLNGKHQTDSIVCEMKMDTDQNTRENLTGINSTVEEPVSPMLPPSAVEEREAVSKTALA
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
CL0381
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Stem Cell Markers, Transcription Factors and Regulators
PBS (pH 7.2) and 40% Glycerol
5978
IgG1
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.