Learn More
NRSF Antibody (CL0381) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-42048UV
Product Code. 30293085
  • £558.00 / 0.10mL
Description
NRSF Monoclonal antibody specifically detects NRSF in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NRSF
Monoclonal
DyLight 350
Neural-restrictive silencer factor, NRSFneuron restrictive silencer factor, RE1-silencing transcription factor, X2 box repressor, XBRrepressor binding to the X2 box
This antibody was developed using a recombinant protein derived from Q13127, with the exact immunogen sequence remaining proprietary.
0.1 mL
Stem Cell Markers, Transcription Factors and Regulators
5978
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL0381
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified