missing translation for 'onlineSavingsMsg'
Learn More

NRK1 Antibody, Novus Biologicals™

Product Code. 18377939 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377939 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377939 Supplier Novus Biologicals Supplier No. H00054981D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NRK1 Polyclonal antibody specifically detects NRK1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRK1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_060351.1
Gene Alias bA235O14.2, C9orf95, cDNA FLJ39825, EC 2.7.1.173, EC 2.7.1.22, EC 2.7.1.n4, FLJ20559, nicotinamide riboside kinase 1, Nicotinic acid riboside kinase 1, NmR-K 1, NMRK1, NRK 1, NRK1, Ribosylnicotinamide kinase 1, Ribosylnicotinic acid kinase 1, RNK 1
Host Species Rabbit
Immunogen C9orf95 (NP_060351.1, 1 a.a. - 199 a.a.) full-length human protein. MKTFIIGISGVTNSGKTTLAKNLQKHLPNCSVISQDDFFKPESEIETDKNGFLQYDVLEALNMEKMMSAISCWMESARHSVVSTDQESAEEIPILIIEGFLLFNYKPLDTIWNRSYFLTIPYEECKRRRSTRVYQPPDSPGYFDGHVWPMYLKYRQEMQDITWEVVYLDGTKSEEDLFLQVYEDLIQELAKQKCLQVTA
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 54981
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.