missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NRIP3 Monoclonal Antibody (OTI1A9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526014
This item is not returnable.
View return policy
Description
NRIP3 Monoclonal Antibody for Western Blot, IHC (P)
NRIP3 is a protein coding gene.
Specifications
| NRIP3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NQ35 | |
| Nrip3 | |
| Full length human recombinant protein of NRIP3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1A9 | |
| Unconjugated | |
| Nrip3 | |
| A330103B05Rik; C11orf14; D7H11orf14; ICRFP703B1614Q5.2; ICRFP703N2430Q5.2; NRIP3; nuclear receptor interacting protein 3; nuclear receptor-interacting protein 3; NY-SAR-105; Sarcoma antigen NY-SAR-105 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 56675 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction