missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NRIP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589510
This item is not returnable.
View return policy
Description
Positive Samples: U-87MG, HT-29, 293T, Mouse brain, Rat brain; Cellular Location: Nucleus Immunogen sequence: QDLIHGQESR RKTSRTEIPA LLVNCKCQDQ LLRVAVDTGT QYNRISAGCL SRLGLEKRVL KASAGDLAPG PPTQVEQLEL QLGQETVVCS AQVVDAESPE FCLGLQTLLS LKCCIDLEHG VLRLKAPFSE LPFLPLYQEP GQ.
NRIP2 down-regulates transcriptional activation by nuclear receptors such as NR1F2.
Specifications
| NRIP2 | |
| Polyclonal | |
| Unconjugated | |
| NRIP2 | |
| AW491344; DKFZP761G1913; fj77f06; neuronal interacting factor 1; neuronal-interacting factor X 1; Nix1; Nrip2; nuclear receptor interacting protein 2; nuclear receptor interacting protein 2-like; nuclear receptor-interacting protein 2; si:dkey-245f7.1; similar to nuclear receptor interacting protein 2; wu:fj77f06 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 60345, 689619, 83714 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.51 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q9BQI9, Q9JHR9 | |
| NRIP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 140-281 of human NRIP2 (NP_1136621). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction