missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRIP Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30075AF532
This item is not returnable.
View return policy
Description
NRIP Polyclonal specifically detects NRIP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NRIP | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| 1200006M05Rik, Androgen receptor complex-associated protein, ARCAP, DDB1 and CUL4 associated factor 6, DDB1- and CUL4-associated factor 6, FLJ23798, IQ motif and WD repeat-containing protein 1, IQ motif and WD repeats 1, IQWD1, MSTP055, NRIP, Nuclear receptor interaction protein, PC326 | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| DCAF6 | |
| Maps to a region between residue 300 and 350 of human nuclear receptor interaction protein (IQ motif and WD repeats 1) using the numbering given in entry NP_001017977.1 | |
| 0.1 mL | |
| 55827.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction