missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRIF3 Antibody (OTI4H1), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73064H
This item is not returnable.
View return policy
Description
NRIF3 Monoclonal specifically detects NRIF3 in Human samples. It is validated for Western Blot.
Specifications
| NRIF3 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| ITGB3BP | |
| Full length recombinant protein of human NRIF3 (NP_055103) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI4H1 | |
| Western Blot | |
| beta-3-endonexin, CENP-R, centromere protein R, HSU37139, integrin beta 3 binding protein (beta3-endonexin), Integrin beta-3-binding protein, NRIF3CENPRbeta 3 endonexin, Nuclear receptor-interacting factor 3, TAP20 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis | |
| 23421.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction