missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRIF3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94407-0.1ml
This item is not returnable.
View return policy
Description
NRIF3 Polyclonal antibody specifically detects NRIF3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NRIF3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| beta-3-endonexin, CENP-R, centromere protein R, HSU37139, integrin beta 3 binding protein (beta3-endonexin), Integrin beta-3-binding protein, NRIF3CENPRbeta 3 endonexin, Nuclear receptor-interacting factor 3, TAP20 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-111 of human ITGB3BP (NP_055103.3). MPVKRSLKLDGLLEENSFDPSKITRKKSVITYSPTTGTCQMSLFASPTSSEEQKHRNGLSNEKRKKLNHPSLTESKESTTKDNDEFMMLLSKVEKLSEEIMEIMQNLSSIQ | |
| 0.1 mL | |
| Apoptosis | |
| 23421 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction