missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NRGN Recombinant Rabbit Monoclonal Antibody (4D4V2)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558045

 View more versions of this product

Product Code. 30282381

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: DCCTENACSK PDDDILDIPL DDPGANAAAA KIQASFRGHM ARKKIKSGER GRKGPGPGGP GGAGVARGGA GGGPSGD.

Neurogranin is the human homolog of the neuron-specific rat RC3/neurogranin gene. This gene encodes a postsynaptic protein kinase substrate that binds calmodulin in the absence of calcium. The NRGN gene contains four exons and three introns. The exons 1 and 2 encode the protein and exons 3 and 4 contain untranslated sequences. It is suggested that the NRGN is a direct target for thyroid hormone in human brain, and that control of expression of this gene could underlay many of the consequences of hypothyroidism on mental states during development as well as in adult subjects.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NRGN
Recombinant Monoclonal
1.35 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P60761, Q04940
NRGN
Recombinant protein (or fragment).
20 μL
Primary
Mouse, Rat
Antibody
IgG
ELISA, Western Blot
4D4V2
Unconjugated
NRGN
0710001B06Rik; AI838505; B-50 immunoreactive C-kinase substrate; BICKS; calmodulin-binding protein; hng; NEUG(55-78); Neurogranin; neurogranin (protein kinase C substrate, RC3); ng; NG/RC3; NRGN; p17; postsynaptic spine 1; protein kinase C substrate; protein kinase C substrate 7.5 kDa protein; protein kinase C substrate, RC3; Pss1; R75334; RC3; similar to neurogranin
Rabbit
Protein A
RUO
64011, 64356
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.