missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRG4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Description
NRG4 Polyclonal antibody specifically detects NRG4 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | NRG4 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:100-1:500 |
| Formulation | 0.2μm filtered solution in PBS without Preservative |
| Gene Alias | DKFZp779N0541, DKFZp779N1944, heregulin 4, HRG4pro-NRG4, neuregulin 4, pro-neuregulin-4, membrane-bound isoform, Pro-NRG4 |
| Host Species | Rabbit |
| Immunogen | Purified, recombinant Human NRG4 (Q8WWG1; Pro2-Phe62) |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?