missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRG4 Antibody (001), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90070AF532
This item is not returnable.
View return policy
Description
NRG4 Monoclonal specifically detects NRG4 in Human samples. It is validated for ELISA.
Specifications
| NRG4 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 145957 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 1 | |
| ELISA | |
| DKFZp779N0541, DKFZp779N1944, heregulin 4, HRG4pro-NRG4, neuregulin 4, pro-neuregulin-4, membrane-bound isoform, Pro-NRG4 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human NRG4 (Q8WWG1; Pro2-Phe62). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction