missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NRG3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542602
This item is not returnable.
View return policy
Description
Peptide sequence: TSINMQLPSR ETNPYFNSLE QKDLVGYSST RASSVPIIPS VGLEETCLQM Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
This gene is a member of the neuregulin gene family. This gene family encodes ligands for the transmembrane tyrosine kinase receptors ERBB3 and ERBB4 - members of the epidermal growth factor receptor family. Ligand binding activates intracellular signaling cascades and the induction of cellular responses including proliferation, migration, differentiation, and survival or apoptosis. This gene encodes neuregulin 3. NRG3 has been shown to activate the tyrosine phosphorylation of its cognate receptor, ERBB4, and is thought to influence neuroblast proliferation, migration and differentiation by signalling through ERBB4. NRG3 also promotes mammary differentiation during embryogenesis. Linkage studies have implicated this gene as a susceptibility locus for schizophrenia and schizoaffective disorder. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Additional transcript variants have been described but their biological validity has not been verified.
Specifications
| NRG3 | |
| Polyclonal | |
| Unconjugated | |
| NRG3 | |
| HGNC:7999; HRG3; neuregulin 3; neuregulin 3 variant 1; neuregulin 3 variant 10; neuregulin 3 variant 11; neuregulin 3 variant 12; neuregulin 3 variant 13; neuregulin 3 variant 14; neuregulin 3 variant 3; neuregulin 3 variant 4; neuregulin 3 variant 5; neuregulin 3 variant 6; neuregulin 3 variant 7; neuregulin 3 variant 8; neuregulin 3 variant 9; Neuregulin-3; neuregulin-3-like polypeptide; NRG3; NRG-3; Pro-neuregulin-3, membrane-bound isoform; pro-NRG3; RGD1559678; scaramanga; ska | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10718 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P56975 | |
| NRG3 | |
| Synthetic peptide directed towards the middle region of human NRG3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction