missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NRG2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA565487

Product Code. 15928234

  • £475.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: HVIRRETETTF SGSHSCSPSH HCSTATPTSS HRHESHTWSL ERSESLTSDS QSGIMLSSVG TSKCNSPACV EARARRAAAY NLEERRRATA PPYHDSVDSL Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.

Neuregulin 2 is a novel member of the neuregulin family of growth and differentiation factors. Through interaction with the Erbb family of receptors, NRG2 induces the growth and differentiation of epithelial, neuronal, glial, and other types of cells. The gene consists of 12 exons and the genomic structure is similar to that of neuregulin 1, another member of the neuregulin family of ligands. NRG1 and NRG2 mediate distinct biological processes by acting at different sites in tissues and eliciting different biological responses in cells. The gene is located close to the region for demyelinating Charcot-Marie-Tooth disease locus, but is not responsible for this disease. Alternative transcripts encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NRG2
Polyclonal
Unconjugated
NRG2
divergent of neuregulin-1; Don1; DON-1; HRG2; neural- and thymus-derived activator for ErbB kinases; neuregulin 2; Neuregulin-2; NRG2; NRG-2; NTAK; NTAK alpha1; NTAK_alpha2a; pro-neuregulin-2, membrane-bound isoform; Pro-NRG2
Rabbit
Antigen Affinity Chromatography
RUO
9542
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.04 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O14511
NRG2
Recombinant Human NRG2. Recombinant protein control fragment (Product #RP-104841).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.