missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NRG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534648
This item is not returnable.
View return policy
Description
Recommended positive controls: Human brain, Mouse brain, rat brain. Predicted reactivity: Rat (97%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Neuregulin 1 (NRG1) was originally identified as a 44-kD glycoprotein that interacts with the NEU/ERBB2 receptor tyrosine kinase to increase its phosphorylation on tyrosine residues. It is known that an extraordinary variety of different isoforms are produced from the NRG1 gene by alternative splicing. These isoforms include heregulins (HRGs), glial growth factors (GGFs) and sensory and motor neuron-derived factor (SMDF). They are tissue-specifically expressed and differ significantly in their structure. The HRG isoforms all contain immunoglobulin (Ig) and epidermal growth factor-like (EGF-like) domains. GGF and GGF2 isoforms contain a kringle-like sequence plus Ig and EGF-like domains; and the SMDF isoform shares only the EGF-like domain with other isoforms. The receptors for all NRG1 isoforms are the ERBB family of tyrosine kinase transmembrane receptors. Through interaction with ERBB receptors, NRG1 isoforms induce the growth and differentiation of epithelial, neuronal, glial, and other types of cells.
Specifications
| NRG1 | |
| Polyclonal | |
| Unconjugated | |
| NRG1 | |
| 6030402G23Rik; Acetylcholine receptor-inducing activity; ARIA; Breast cancer cell differentiation factor p45; D230005F13Rik; GGF; GGF2; GGFII; Glial growth factor; glial growth factor 2; heregulin; heregulin, alpha (45kD, ERBB2 p185-activator); HGL; HRG; HRG1; HRGA; HRGalpha; MST131; MSTP131; NDF; Neu differentiation factor; neuregulin 1; neuregulin 1 type III beta 3; Neuregulin-1; NRG1; NRG-1; NRG1-IT2; pro-neuregulin-1, membrane-bound isoform; Pro-NRG1; sensory and motor neuron derived factor; Sensory and motor neuron-derived factor; SMDF; type I neuregulin 1; type III neuregulin 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 112400, 211323, 3084 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.96 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P43322, Q02297 | |
| NRG1 | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human Neuregulin-1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction