missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRG1 Monoclonal Antibody (7D5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512896
This item is not returnable.
View return policy
Description
MA5-12896 targets Heregulin in immunohistochemistry (paraffin), immunoprecipitation, and Western blot applications and shows reactivity with Human, mouse, and Rat samples. The MA5-12896 immunogen is recombinant extracellular domain of rat NDF protein.
NRG1 Monoclonal antibody specifically detects NRG1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Specifications
| NRG1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P43322, Q02297 | |
| NRG1 | |
| Recombinant extracellular domain of rat NDF protein | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 7D5 | |
| Unconjugated | |
| NRG1 | |
| 6030402G23Rik; Acetylcholine receptor-inducing activity; ARIA; Breast cancer cell differentiation factor p45; D230005F13Rik; GGF; GGF2; GGFII; Glial growth factor; glial growth factor 2; heregulin; heregulin, alpha (45kD, ERBB2 p185-activator); HGL; HRG; HRG1; HRGA; HRGalpha; MST131; MSTP131; NDF; Neu differentiation factor; neuregulin 1; neuregulin 1 type III beta 3; Neuregulin-1; NRG1; NRG-1; NRG1-IT2; pro-neuregulin-1, membrane-bound isoform; Pro-NRG1; sensory and motor neuron derived factor; Sensory and motor neuron-derived factor; SMDF; type I neuregulin 1; type III neuregulin 1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 112400, 211323, 3084 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction