missing translation for 'onlineSavingsMsg'
Learn More

NRF3 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18384385 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384385 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384385 Supplier Novus Biologicals Supplier No. NBP305800FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NRF3 Polyclonal specifically detects NRF3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRF3
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias NF-E2-related factor 3, NFE2-related factor 3, Nrf3, NRF3nuclear factor erythroid 2-related factor 3, nuclear factor (erythroid-derived 2)-like 3, Nuclear factor, erythroid derived 2, like 3, nuclear factor-erythroid 2 p45-related factor 3, nuclear factor-erythroid 2-related factor 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NRF3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9603
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.