missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nrf2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA5142785
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize all reported isoforms. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Nfr1 (also designated TCF11), Nrf2 and Nfr3 are bZIP transcription factors that heterodimer with Maf proteins to bind MARE (Maf recognition elements) sequences. Nrf2 is widely expressed and is thought to translocate to the nucleus after treatment with xenobiotics and antioxidants, which stimulate its release from a repressor protein Keap1.Specifications
Nrf2 | |
Polyclonal | |
Unconjugated | |
NFE2L2 | |
AI194320; E4TF1-60; E4TF1A; HEBP1; Nfe2l2; NF-E2-related factor 2; NFE2-related factor 2; NF-E2-related factor-2; NFT2; NRF2; NRF2A; nuclear factor (erythroid-derived 2)-like 2; nuclear factor erythroid 2-like 2; nuclear factor erythroid 2-related factor 2; nuclear factor erythroid-derived 2-like 2; nuclear factor, erythroid 2 like 2; nuclear factor, erythroid 2-like 2; nuclear factor, erythroid derived 2, like 2; Unknown (protein for IMAGE:7945830) | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
4780 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
Flow Cytometry, Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q16236 | |
NFE2L2 | |
Peptide with sequence C-KHSSRLEAHLTRDE. | |
100 μg | |
Primary | |
Human | |
Liquid |