Learn More
Invitrogen™ Nrf2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120048
Description
Immunogen sequence: SPHFLNAFED SFSSILSTED PNQLTVNSLN SDATVNTDFG DEFYSAFIAE PSISNSMPSP ATLSHSLSEL LNGPIDVSDL SLCKAFNQNH PESTAEFNDS DSGISLNTSP SVASPEHSVE SSSYGDTLLG LSDSEVEELD SAPGSVKQNG PKTPVHSS Positive test controls include: HeLa, NIH/3T3. The target is usually found in the following locations: Cytoplasm, Nucleus, cytosol.
Nfr1 (also designated TCF11), Nrf2 and Nfr3 are bZIP transcription factors that heterodimer with Maf proteins to bind MARE (Maf recognition elements) sequences. Nrf2 is widely expressed and is thought to translocate to the nucleus after treatment with xenobiotics and antioxidants, which stimulate its release from a repressor protein Keap1.
Specifications
| Nrf2 | |
| Polyclonal | |
| 0.92 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O54968, Q16236, Q60795 | |
| NFE2L2 | |
| Recombinant Protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 6W6R7 | |
| Unconjugated | |
| NFE2L2 | |
| AI194320; E4TF1-60; E4TF1A; HEBP1; Nfe2l2; NF-E2-related factor 2; NFE2-related factor 2; NF-E2-related factor-2; NFT2; NRF2; NRF2A; nuclear factor (erythroid-derived 2)-like 2; nuclear factor erythroid 2-like 2; nuclear factor erythroid 2-related factor 2; nuclear factor erythroid-derived 2-like 2; nuclear factor, erythroid 2 like 2; nuclear factor, erythroid 2-like 2; nuclear factor, erythroid derived 2, like 2; Unknown (protein for IMAGE:7945830) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18024, 4780, 83619 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.