missing translation for 'onlineSavingsMsg'
Learn More

Nrf2 Antibody (OTI8A10) [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30688804 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30688804 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688804 Supplier Novus Biologicals Supplier No. NBP344685MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nrf2 Monoclonal antibody specifically detects Nrf2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nrf2
Applications Western Blot
Classification Monoclonal
Clone OTI8A10
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias HEBP1, NF-E2-related factor 2, NFE2-related factor 2, NRF2 erythroid derived 2, like 2, nuclear factor (erythroid-derived 2)-like 2, nuclear factor erythroid 2-related factor 2, nuclear factor erythroid-derived 2-like 2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 330-605 of human NFE2L2 (NP_006155) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4780
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.