missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nrf1 Rabbit anti-Human, Mouse, Rat, Clone: 4Q4E7, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £386.00
Specifications
| Antigen | Nrf1 |
|---|---|
| Clone | 4Q4E7 |
| Dilution | Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin |
| Applications | ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18399395
|
Novus Biologicals
NBP3-16161-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18340974
|
Novus Biologicals
NBP3-16161-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nrf1 Monoclonal antibody specifically detects Nrf1 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| Nrf1 | |
| Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Alpha palindromic-binding protein, ALPHA-PAL, EWG, NRF-1, nuclear respiratory factor 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 300-400 of human Nrf1 (Q16656). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 4Q4E7 | |
| ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Neurodegeneration, Neuroscience | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4899 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title