Learn More
NRF1/nuclear respiratory factor 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12482-1-AP-150UL
 View more versions of this product
Product Code. 16800224
  • £349.00 / 150µL
Description
This gene encodes a protein that homodimerizes and functions as a transcription factor which activates the expression of some key metabolic genes regulating cellular growth and nuclear genes required for respiration, heme biosynthesis, and mitochondrial DNA transcription and replication. The protein has also been associated with the regulation of neurite outgrowth. Alternate transcriptional splice variants, which encode the same protein, have been characterized. Additional variants encoding different protein isoforms have been described but they have not been fully characterized. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene and for ""nuclear factor (erythroid-derived 2)-like 1"" which has an official symbol of NFE2L1.
Specifications
Specifications
NRF1/nuclear respiratory factor 1
Polyclonal
Unconjugated
NRF1
ALPHA PAL, NRF 1, NRF1, NRF1/nuclear respiratory factor 1, nuclear respiratory factor 1
Rabbit
Antigen Affinity Chromatography
RUO
18181, 312195, 4899
-20°C
Liquid
Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q16656, Q62792, Q9WU00
Nrf1
NRF1/nuclear respiratory factor 1 Fusion Protein Ag3172
150 μL
Primary
Human, Mouse, Rat