Learn More
Invitrogen™ NRD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110042
Description
Immunogen sequence: SGAEIEDDDE EGFDDEDEFD DEHDDDLDTE DNELEELEER AEARKKTTEK QSAAALCVGV GSFADPDDLP GLAHFLEHMV FMGSLKYPDE NGFDAFLKKH GGSDNASTDC ERTVFQFDVQ RKYFKEALDR WAQFFIHPLM IRDAIDREVE AVDSEYQLAR PSDANRKEML FGSLARPGHP MGKFFWGNAE TLKHEPRKNN IDTHARLREF WMRYYSSHYM TLVVQSKETL DTLEKWVTEI F.
Cleaves peptide substrates on the N-terminus of arginine residues in dibasic pairs.
Specifications
| NRD1 | |
| Polyclonal | |
| Unconjugated | |
| Nrdc | |
| 2600011I06Rik; AI875733; hNRD1; hNRD2; nardilysin; nardilysin (N-arginine dibasic convertase); nardilysin 1; nardilysin 1 (N-arginine dibasic convertase); nardilysin convertase; nardilysin, N-arginine dibasic convertase 1; nardilysin, N-arginine dibasic convertase, NRD convertase 1; N-arginine dibasic convertase; N-arginine dibasic convertase 1; NRD convertase; Nrd1; NRDC; NRD-C | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 230598, 25499, 4898 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.63 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43847, P47245, Q8BHG1 | |
| Nrd1, Nrdc | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 160-400 of human NRD1 (NP_001095132.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.